Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000038.5 | NP_000029.2 | 2 | 169 | ||
NM_001127510.2 | NP_001120982.1 | 3 | 277 | |||
GenBank mRNA | AB210001.1 | - | 3 | 241 | ||
BC056268.1 | - | 2 | 140 | |||
DA777787.1 | - | 2 | 464 | |||
DA795663.1 | - | 2 | 169 | |||
M73548.1 | - | 1 | 122 | |||
M74088.1 | - | 1 | 102 |