Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000511.5 | NP_000502.4 | 2 | 949 | ||
NM_001097638.2 | NP_001091107.1 | 2 | 939 | |||
GenBank mRNA | AK290277.1 | - | 2 | 935 | ||
AY751739.1 | - | 1 | 828 | |||
BC001899.1 | - | 2 | 889 | |||
BC121066.1 | - | 2 | 871 | |||
D87942.1 | - | 2 | 939 |