Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000608.2 | NP_000599.1 | 3 | 385 | ||
GenBank mRNA | AK303760.1 | - | 2 | 717 | ||
AK312226.1 | - | 3 | 305 | |||
AV661485.1 | - | 3 | 385 | |||
BC015964.1 | - | 3 | 309 | 3934986 | ||
BC056239.1 | - | 3 | 309 | 6463124 | ||
CR456930.1 | - | 3 | 269 |