Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002105.2 | NP_002096.1 | 1 | 935 | ||
GenBank mRNA | BC004915.2 | - | 1 | 905 | 3537648 | |
BC011694.2 | - | 1 | 915 | 3139343 | ||
BC013416.2 | - | 1 | 900 | 3534359 | ||
X14850.1 | - | 1 | 934 |