Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320605.1 | NP_001307534.1 | 2 | 367 | ||
NM_001320606.1 | NP_001307535.1 | 2 | 664 | |||
NM_001320608.1 | NP_001307537.1 | 2 | 367 | |||
NM_002162.4 | NP_002153.2 | 2 | 367 | |||
GenBank mRNA | AF340060.1 | - | 2 | 120 | ||
AF340061.1 | - | 2 | 120 | |||
AK226023.1 | - | 2 | 201 | |||
AK309643.1 | - | 2 | 449 | |||
AK312826.1 | - | 2 | 202 | |||
AX746321.1 | - | 2 | 162 | |||
BC040123.1 | - | 2 | 147 | |||
BC046121.1 | - | 2 | 194 | |||
BC058903.1 | - | 2 | 153 | 5207143 | ||
CR982084.1 | - | 2 | 367 | |||
DC425414.1 | - | 2 | 356 | |||
S50015.1 | - | 2 | 148 | |||
X69711.1 | - | 2 | 162 | |||
X69819.1 | - | 2 | 155 |