Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002173.3 | NP_002164.1 | 1 | 240 | ||
GenBank mRNA | M28585.1 | - | 1 | 240 | ||
X00140.1 | - | 1 | 240 |