Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002982.3 | NP_002973.1 | 1, 2 | 153 | ||
GenBank mRNA | AK311960.1 | - | 1, 2 | 145 | ||
AV733621.2 | - | 1, 2 | 153 | |||
BC009716.1 | - | 1, 2 | 136 | 3901489 | ||
BG530064.1 | - | 1, 2 | 168 | |||
BT007329.1 | - | 1, 2 | 80 | |||
M24545.1 | - | 1, 2 | 149 | |||
M28226.1 | - | 1, 2 | 126 | |||
S69738.1 | - | 1, 2 | 111 | |||
S71513.1 | - | 1, 2 | 133 | |||
X14768.1 | - | 1, 2 | 133 |