Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000796.5 | NP_000787.2 | 3 | 722 | ||
NM_001282563.2 | NP_001269492.1 | 4 | 581 | |||
NM_001290809.1 | NP_001277738.1 | 4 | 714 | |||
NM_033663.5 | NP_387512.3 | 3 | 722 | |||
XM_017005829.1 | XP_016861318.1 | 4 | 542 | |||
GenBank mRNA | AK292309.1 | - | 4 | 581 | ||
BC095510.1 | - | 2 | 292 | 7001986 | ||
BC128123.1 | - | 3 | 368 | 40112300 | ||
KJ534829.1 | - | 2 | 291 | |||
L20469.1 | - | 2 | 291 | |||
U32499.1 | - | 2 | 291 |
Set Membership: |
G-protein coupled receptor |