Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000871.2 | NP_000862.1 | 3 | 1894 | ||
GenBank mRNA | AF007141.1 | - | 1 | 103 | ||
L41147.1 | - | 3 | 1894 |
Set Membership: |
G-protein coupled receptor |