Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313905.1 | NP_001300834.1 | 3 | 183 | ||
NM_001425.2 | NP_001416.1 | 3 | 367 | |||
XM_011526605.2 | XP_011524907.1 | 4 | 298 | |||
GenBank mRNA | AL541088.3 | - | 3 | 367 | ||
BC009718.1 | - | 3 | 174 | 3891050 | ||
BT007124.1 | - | 2 | 113 | |||
CR456871.1 | - | 2 | 113 | |||
CR541955.1 | - | 2 | 113 | |||
U52101.1 | - | 3 | 162 | |||
U87947.1 | - | 3 | 354 | |||
X94771.1 | - | 2 | 116 |