Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204406.1 | NP_001191335.1 | 3 | 515 | ||
NM_001629.3 | NP_001620.2 | 2 | 194 | |||
GenBank mRNA | AK314143.1 | - | 3 | 515 | ||
BC018538.1 | - | 2 | 147 | 4342203 | ||
BG569867.1 | - | 2 | 196 |