Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001700.4 | NP_001691.1 | 2, 3 | 229 | ||
GenBank mRNA | BC044940.1 | - | 2, 3 | 251 | ||
BC069495.1 | - | 2, 3 | 224 | 7262247 | ||
BC093931.1 | - | 2, 3 | 229 | 7939776 | ||
BC093933.1 | - | 2, 3 | 229 | 7939778 | ||
X58794.1 | - | 2, 3 | 202 |