Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277163.2 | NP_001264092.1 | 5 | 719 | ||
GenBank mRNA | L00693.1 | - | 5 | 685 |