Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002411.3 | NP_002402.1 | 2 | 212 | ||
XM_005274005.3 | XP_005274062.3 | 2 | 237 | |||
GenBank mRNA | AI904278.1 | - | 2 | 212 | ||
AY217100.1 | - | 2 | 150 | |||
BC067220.1 | - | 2 | 213 | 30321681 | ||
BC128252.1 | - | 2 | 204 | 40115103 | ||
BC128402.1 | - | 2 | 204 | 40115099 | ||
U33147.1 | - | 2 | 210 |