Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003225.2 | NP_003216.1 | 1, 2 | 129 | ||
GenBank mRNA | BC032811.1 | - | 1, 2 | 113 | 5186922 | |
BQ083523.1 | - | 1, 2 | 188 | |||
M12075.1 | - | 1, 2 | 88 | |||
X00474.1 | - | 1, 2 | 129 | |||
X52003.1 | - | 1, 2 | 129 |