Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003695.2 | NP_003686.1 | 2, 3 | 222 | ||
GenBank mRNA | AK312164.1 | - | 2, 3 | 169 | ||
BC022806.1 | - | 2, 3 | 149 | |||
BC031330.1 | - | 2, 3 | 222 | 5428818 | ||
BM019592.1 | - | 2, 3 | 233 | |||
X82693.1 | - | 2, 3 | 157 |