Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004647.3 | NP_004638.2 | 6, 7 | 695 | ||
GenBank mRNA | U43843.1 | - | 6, 7 | 593 |