Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004720.5 | NP_004711.2 | 2 | 860 | ||
XM_011528421.2 | XP_011526723.1 | 2 | 1224 | |||
XM_017027468.1 | XP_016882957.1 | 4 | 1155 | |||
XM_017027469.1 | XP_016882958.1 | 3 | 958 | |||
GenBank mRNA | AF011466.1 | - | 2 | 805 | ||
AF197929.1 | - | 2 | 839 | |||
AF233092.1 | - | 1 | 727 | |||
AK123043.1 | - | 5 | 1624 | |||
AK316573.1 | - | 2 | 821 | |||
AY322548.1 | - | 1 | 721 | |||
BC025695.1 | - | 2 | 900 | 5209267 |
Set Membership: |
G-protein coupled receptor |