Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAL3ST1 SIRNA Assays ›
Gene Symbol:
GAL3ST1
Gene Name:
galactose-3-O-sulfotransferase 1
Gene Aliases:
CST
Chromosome Location:
Chr.22: 30554635 - 30574588 on Build GRCh38
UniGene ID:
Hs.17958
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
45004
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GAL3ST1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318103.1
NP_001305032.1
3
652
NM_001318104.1
NP_001305033.1
4
762
NM_001318105.1
NP_001305034.1
3
713
NM_001318106.1
NP_001305035.1
4
838
NM_001318107.1
NP_001305036.1
4
826
NM_001318108.1
NP_001305037.1
4
823
NM_001318109.1
NP_001305038.1
3
597
NM_001318110.1
NP_001305039.1
4
730
NM_001318111.1
NP_001305040.1
3
620
NM_001318112.1
NP_001305041.1
3
566
NM_001318113.1
NP_001305042.1
4
676
NM_001318114.1
NP_001305043.1
3
569
NM_001318115.1
NP_001305044.1
4
695
NM_001318116.1
NP_001305045.1
5
818
NM_004861.2
NP_004852.1
4
749
XM_011530518.2
XP_011528820.1
3
2187
XM_011530522.2
XP_011528824.1
3
2303
XM_011530524.1
XP_011528826.1
3
656
XM_011530528.1
XP_011528830.1
3
557
XM_017029096.1
XP_016884585.1
3
773
XM_017029097.1
XP_016884586.1
3
2071
XM_017029098.1
XP_016884587.1
4
2196
GenBank mRNA
AK313097.1
-
3
674
BC014649.1
-
4
656
4564703
BC019077.2
-
3
551
4562507
BG419785.1
-
4
673
BP276494.1
-
3
566
CR456350.1
-
3
594
D88667.1
-
4
724
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
protein N-linked glycosylation
galactosylceramide biosynthetic process
spermatogenesis
myelination
galactosylceramide sulfotransferase activity
sulfotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0