Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004995.3 | NP_004986.1 | 1, 2 | 402 | ||
GenBank mRNA | BC064803.1 | - | 1, 2 | 336 | 6046773 | |
D26512.1 | - | 1, 2 | 228 | |||
HE978355.1 | - | 1, 2 | 351 | |||
HY075830.1 | - | 1, 2 | 402 | |||
U41078.1 | - | 1, 2 | 117 | |||
X83535.1 | - | 1, 2 | 236 | |||
X90925.1 | - | 1, 2 | 229 | |||
Z48481.1 | - | 1, 2 | 230 |