Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178078.1 | NP_001171549.1 | 4 | 646 | ||
NM_001178079.1 | NP_001171550.1 | 4 | 572 | |||
NM_001178080.1 | NP_001171551.1 | 3 | 490 | |||
NM_001178081.1 | NP_001171552.1 | 2 | 351 | |||
NM_003153.4 | NP_003144.3 | 4 | 627 | |||
NR_033659.1 | 3 | 488 | ||||
XM_006719575.2 | XP_006719638.1 | 5 | 624 | |||
XM_011538703.2 | XP_011537005.1 | 4 | 647 | |||
XM_011538704.2 | XP_011537006.1 | 5 | 658 | |||
XM_011538705.2 | XP_011537007.1 | 5 | 768 | |||
XM_011538706.2 | XP_011537008.1 | 5 | 624 | |||
XM_011538707.2 | XP_011537009.1 | 4 | 609 | |||
XM_011538708.2 | XP_011537010.1 | 3 | 451 | |||
XM_011538709.2 | XP_011537011.1 | 4 | 485 | |||
GenBank mRNA | AK290431.1 | - | 4 | 575 | ||
AK291044.1 | - | 4 | 573 | |||
AK298721.1 | - | 2 | 171 | |||
AK299094.1 | - | 3 | 424 | |||
AK308751.1 | - | 4 | 470 | |||
AK312471.1 | - | 4 | 493 | |||
AK316142.1 | - | 3 | 434 | |||
AK316236.1 | - | 3 | 265 | |||
BC075852.1 | - | 4 | 428 | 5530399 | ||
BT007380.1 | - | 3 | 321 | |||
U16031.1 | - | 4 | 486 |