Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005105.4 | NP_005096.1 | 6 | 662 | ||
GenBank mRNA | AF127761.1 | - | 6 | 564 | ||
AF161463.1 | - | 6 | 576 | |||
AF182415.1 | - | 6 | 581 | |||
AF198620.1 | - | 6 | 583 | |||
AF231511.1 | - | 6 | 569 | |||
AK075009.1 | - | 6 | 583 | |||
BC017088.2 | - | 6 | 624 | 3850355 | ||
BC071577.1 | - | 6 | 610 |