Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005345.5 | NP_005336.3 | 1 | 2211 | ||
GenBank mRNA | AB209818.1 | - | 1 | 2172 | ||
AK096017.1 | - | 2 | 1687 | |||
AK225692.1 | - | 1 | 2182 | |||
AK225699.1 | - | 1 | 2182 | |||
AK291295.1 | - | 1 | 2182 | |||
AK294182.1 | - | 2 | 1975 | |||
AK298056.1 | - | 2 | 2110 | |||
AK298098.1 | - | 2 | 1510 | |||
AK301243.1 | - | 2 | 1891 | |||
AK301574.1 | - | 2 | 2128 | |||
AK303980.1 | - | 2 | 1453 | |||
AK304618.1 | - | 2 | 1948 | |||
AK304652.1 | - | 2 | 2017 | |||
BC002453.2 | - | 1 | 2145 | 3345864 | ||
FJ224292.1 | - | 1 | 2457 |