Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307980.1 | NP_001294909.1 | 3 | 749 | ||
NM_001307981.1 | NP_001294910.1 | 3 | 749 | |||
NM_005528.3 | NP_005519.2 | 4 | 788 | |||
GenBank mRNA | AF012106.1 | - | 1 | 963 | ||
AF036874.1 | - | 3 | 395 | |||
BC032137.2 | - | 3 | 423 | |||
BC044584.1 | - | 4 | 791 | 4811478 | ||
BC051008.1 | - | 3 | 271 | |||
BC053947.1 | - | 4 | 791 | |||
BC057849.1 | - | 3 | 621 | 30349314 |