Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003430.3 | NP_003421.2 | 3 | 392 | ||
XM_011528265.2 | XP_011526567.1 | 3 | 353 | |||
XM_011528266.2 | XP_011526568.1 | 3 | 307 | |||
XM_017027235.1 | XP_016882724.1 | 3 | 346 | |||
XM_017027236.1 | XP_016882725.1 | 2 | 213 | |||
GenBank mRNA | AK291256.1 | - | 3 | 299 | ||
DB068854.1 | - | 3 | 392 | |||
DB219374.1 | - | 3 | 299 | |||
L11672.1 | - | 3 | 280 | |||
M61871.1 | - | 2 | 144 |