Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST2H2AC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003517.2 | NP_003508.1 | 1 | 376 | ||
GenBank mRNA | BC060324.1 | - | 1 | 411 | 3892650 |