Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004158.2 | NP_004149.1 | 2 | 261 | ||
GenBank mRNA | AF040962.1 | - | 2 | 261 | ||
AK055490.1 | - | 1 | 1824 | |||
BC143894.1 | - | 1 | 380 |