Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004292.2 | NP_004283.2 | 4 | 553 | ||
XM_011545399.2 | XP_011543701.1 | 4 | 902 | |||
XM_011545400.2 | XP_011543702.1 | 4 | 458 | |||
XM_011545401.2 | XP_011543703.1 | 4 | 427 | |||
XM_017018587.1 | XP_016874076.1 | 4 | 458 | |||
GenBank mRNA | AB209445.1 | - | 3 | 190 | ||
AK297757.1 | - | 3 | 249 | |||
AK301430.1 | - | 1 | 53 | |||
BC014417.1 | - | 4 | 443 | 4549333 | ||
KU178481.1 | - | 4 | 426 | |||
KU178482.1 | - | 4 | 426 | |||
L36463.1 | - | 4 | 553 |