Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004720.5 | NP_004711.2 | 2 | 870 | ||
XM_011528421.2 | XP_011526723.1 | 2 | 1234 | |||
XM_017027468.1 | XP_016882957.1 | 4 | 1165 | |||
XM_017027469.1 | XP_016882958.1 | 3 | 968 | |||
GenBank mRNA | AF011466.1 | - | 2 | 815 | ||
AF197929.1 | - | 2 | 849 | |||
AF233092.1 | - | 1 | 737 | |||
AK123043.1 | - | 5 | 1634 | |||
AK316573.1 | - | 2 | 831 | |||
AY322548.1 | - | 1 | 731 | |||
BC025695.1 | - | 2 | 910 | 5209267 |
Set Membership: |
G-protein coupled receptor |