Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004995.3 | NP_004986.1 | 2 | 413 | ||
GenBank mRNA | BC064803.1 | - | 2 | 347 | 6046773 | |
D26512.1 | - | 2 | 239 | |||
HE978355.1 | - | 2 | 362 | |||
HY075830.1 | - | 2 | 413 | |||
U41078.1 | - | 2 | 128 | |||
X83535.1 | - | 2 | 247 | |||
X90925.1 | - | 2 | 240 | |||
Z48481.1 | - | 2 | 241 |