Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005281.3 | NP_005272.1 | 2 | 546 | ||
GenBank mRNA | AK291185.1 | - | 2 | 533 | ||
AK314868.1 | - | 2 | 525 | |||
BC032702.1 | - | 2 | 542 | 5247608 | ||
BQ882813.1 | - | 2 | 546 | |||
HQ709189.1 | - | 1 | 447 |
Set Membership: |
G-protein coupled receptor |