Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005423.4 | NP_005414.1 | 2, 3 | 375 | ||
GenBank mRNA | BC032820.1 | - | 2, 3 | 254 | 5184770 | |
BM817851.1 | - | 2, 3 | 274 | |||
BM853946.1 | - | 2, 3 | 272 | |||
X51698.1 | - | 2, 3 | 238 |