Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320454.1 | NP_001307383.1 | 3 | 346 | ||
NM_006365.2 | NP_006356.1 | 5 | 478 | |||
NR_135263.1 | 4 | 382 | ||||
NR_135264.1 | 4 | 382 | ||||
NR_135268.1 | 5 | 712 | ||||
XM_011509070.1 | XP_011507372.1 | 4 | 791 | |||
XM_011509071.2 | XP_011507373.1 | 4 | 1578 | |||
XM_011509072.1 | XP_011507374.1 | 4 | 401 | |||
XM_011509073.2 | XP_011507375.1 | 3 | 565 | |||
XM_011509074.1 | XP_011507376.1 | 3 | 346 | |||
XM_017000061.1 | XP_016855550.1 | 6 | 629 | |||
XM_017000062.1 | XP_016855551.1 | 8 | 959 | |||
XM_017000063.1 | XP_016855552.1 | 6 | 583 | |||
XM_017000065.1 | XP_016855554.1 | 7 | 768 | |||
GenBank mRNA | AK124566.1 | - | 1 | 1089 | ||
BC050448.1 | - | 4 | 247 | |||
BC059943.1 | - | 4 | 233 | |||
BI668594.1 | - | 4 | 265 | |||
DB534652.2 | - | 2 | 157 | |||
HY139418.1 | - | 3 | 346 | |||
HY158402.1 | - | 4 | 260 | |||
U49857.1 | - | 5 | 322 |