Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012727.1 | NP_001012745.1 | 1, 2 | 284 | ||
NM_006412.3 | NP_006403.2 | 1, 2 | 284 | |||
GenBank mRNA | AF000237.1 | - | 1, 2 | 248 | ||
AF011374.1 | - | 1, 2 | 242 | |||
BC000026.2 | - | 1, 2 | 227 | 3507150 | ||
BC004529.2 | - | 1, 2 | 218 | 3347049 | ||
BC019292.1 | - | 1, 2 | 236 | 2823580 | ||
BF347518.1 | - | 1, 2 | 284 | |||
CN407343.1 | - | 1, 2 | 284 | |||
U56418.2 | - | 1, 2 | 221 |