Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278730.1 | NP_001265659.1 | 2 | 291 | ||
NM_006478.4 | NP_006469.2 | 2 | 319 | |||
NM_152236.2 | NP_689422.1 | 2 | 324 | |||
XM_011529824.1 | XP_011528126.1 | 3 | 443 | |||
XM_011529825.1 | XP_011528127.1 | 2 | 252 | |||
XM_017028533.1 | XP_016884022.1 | 2 | 969 | |||
GenBank mRNA | AK093386.1 | - | 2 | 261 | ||
AK122829.1 | - | 1 | 1161 | |||
AK223602.1 | - | 2 | 263 | |||
AK225484.1 | - | 2 | 263 | |||
BC001782.1 | - | 2 | 259 | 3355168 | ||
BC007624.1 | - | 2 | 259 | 3357387 | ||
BC011047.1 | - | 2 | 262 | 4178571 | ||
BI601743.1 | - | 2 | 284 | |||
Y07846.1 | - | 2 | 245 |