Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201550.2 | NP_001188479.1 | 6 | 1112 | ||
NM_001201551.1 | NP_001188480.1 | 6 | 1109 | |||
XM_006711129.3 | XP_006711192.1 | 6 | 1051 | |||
XM_017000110.1 | XP_016855599.1 | 6 | 1075 | |||
XM_017000111.1 | XP_016855600.1 | 6 | 1075 | |||
XM_017000112.1 | XP_016855601.1 | 6 | 1075 | |||
XM_017000113.1 | XP_016855602.1 | 6 | 1075 | |||
XM_017000114.1 | XP_016855603.1 | 3 | 503 | |||
GenBank mRNA | AJ640130.2 | - | 6 | 1041 | ||
AK292752.1 | - | 6 | 1109 | |||
BC074957.2 | - | 3 | 282 | 30915282 | ||
X98337.1 | - | 3 | 319 |