Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILVBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006844.4 | NP_006835.2 | 4 | 564 | ||
XM_005259717.4 | XP_005259774.1 | 4 | 711 | |||
XM_011527651.2 | XP_011525953.1 | 3 | 543 | |||
GenBank mRNA | AB209065.1 | - | 2 | 733 | ||
BC000109.1 | - | 3 | 508 | |||
BC011722.2 | - | 4 | 531 | 4300207 | ||
BC011761.2 | - | 4 | 457 | 3622453 | ||
BC126913.1 | - | 2 | 304 | |||
U61263.1 | - | 4 | 439 |