Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284330.1 | NP_001271259.1 | 4 | 729 | ||
NM_001284331.1 | NP_001271260.1 | 4 | 729 | |||
NM_013284.3 | NP_037416.1 | 4 | 729 | |||
NR_104299.1 | 4 | 729 | ||||
XM_006715692.2 | XP_006715755.1 | 4 | 700 | |||
XM_011515275.1 | XP_011513577.1 | 4 | 715 | |||
XM_011515278.1 | XP_011513580.1 | 4 | 715 | |||
XM_011515279.1 | XP_011513581.1 | 4 | 715 | |||
XM_011515285.1 | XP_011513587.1 | 1 | 73 | |||
XM_011515286.2 | XP_011513588.1 | 4 | 715 | |||
XM_011515287.2 | XP_011513589.1 | 4 | 715 | |||
XM_017011998.1 | XP_016867487.1 | 4 | 715 | |||
XM_017011999.1 | XP_016867488.1 | 4 | 715 | |||
XM_017012000.1 | XP_016867489.1 | 4 | 715 | |||
XM_017012001.1 | XP_016867490.1 | 4 | 715 | |||
XM_017012002.1 | XP_016867491.1 | 4 | 682 | |||
XM_017012004.1 | XP_016867493.1 | 4 | 715 | |||
XM_017012005.1 | XP_016867494.1 | 4 | 715 | |||
GenBank mRNA | AF176097.1 | - | 4 | 669 | ||
AJ131891.2 | - | 4 | 672 | |||
AK023002.1 | - | 4 | 704 | |||
AK291638.1 | - | 4 | 673 | |||
AK294449.1 | - | 3 | 601 | |||
BC026306.1 | - | 4 | 700 | 4813399 | ||
BC049202.1 | - | 4 | 674 | 5246144 | ||
BC062590.1 | - | 4 | 653 | 5215523 |