Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135861.2 | NP_001129333.1 | 1, 2 | 776 | ||
NM_001287343.1 | NP_001274272.1 | 1, 2 | 776 | |||
NM_014172.5 | NP_054891.2 | 1, 2 | 776 | |||
NR_109807.1 | 1, 2 | 776 | ||||
GenBank mRNA | AF131857.1 | - | 1, 2 | 500 | ||
AF161490.1 | - | 1, 2 | 181 | |||
AF164795.1 | - | 1, 2 | 169 | |||
AF285119.1 | - | 1, 2 | 494 | |||
AF393504.1 | - | 1, 2 | 161 | |||
AL136644.1 | - | 1, 2 | 186 | |||
BC024648.1 | - | 1, 2 | 182 | 4934238 | ||
BE962935.2 | - | 1, 2 | 172 | |||
BG681446.1 | - | 1, 2 | 778 | |||
BQ922335.1 | - | 1, 2 | 191 | |||
CD242442.1 | - | 1, 2 | 389 | |||
CD693554.1 | - | 1, 2 | 221 | |||
CN334799.1 | - | 1, 2 | 159 | |||
CR533481.1 | - | 1, 2 | 161 | |||
EU794646.1 | - | 1, 2 | 493 |