Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003223.2 | NP_003214.1 | 4 | 655 | ||
XM_011522633.2 | XP_011520935.1 | 4 | 791 | |||
XM_011522635.2 | XP_011520937.1 | 4 | 471 | |||
GenBank mRNA | AB209146.1 | - | 3 | 4797 | ||
AK309301.1 | - | 3 | 1096 | |||
BC010576.1 | - | 4 | 544 | 4181538 | ||
BU159415.1 | - | 4 | 608 | |||
CR541726.1 | - | 4 | 401 | |||
S73885.1 | - | 4 | 659 | |||
X57435.1 | - | 4 | 350 |