Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIA-RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037775.1 | 3 | 285 | |||
GenBank mRNA | AF087861.1 | - | 2 | 65 | ||
AK302147.1 | - | 4 | 701 |
Gene Symbol: MIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202553.1 | NP_001189482.1 | 4 | 320 | ||
NM_006533.3 | NP_006524.1 | 3 | 445 | |||
GenBank mRNA | BC005910.1 | - | 3 | 304 | 4333499 | |
BM715936.1 | - | 3 | 296 | |||
BM856478.1 | - | 3 | 302 | |||
BQ681978.1 | - | 4 | 320 | |||
BT007044.1 | - | 3 | 279 | |||
BX280685.1 | - | 3 | 445 | |||
CR541648.1 | - | 3 | 279 | |||
X75450.1 | - | 3 | 318 |