Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006645.2 | NP_006636.2 | 4 | 1330 | ||
GenBank mRNA | AF151810.1 | - | 4 | 866 | ||
AK095668.1 | - | 4 | 1246 | |||
AK315255.1 | - | 5 | 1124 | |||
BC007919.2 | - | 5 | 849 | 4301295 | ||
BC014033.2 | - | 5 | 849 | 4330790 | ||
BC035482.1 | - | 4 | 1130 |