Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILVBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006844.4 | NP_006835.2 | 16 | 2114 | ||
XM_005259717.4 | XP_005259774.1 | 16 | 2261 | |||
XM_011527651.2 | XP_011525953.1 | 15 | 2093 | |||
GenBank mRNA | AB209065.1 | - | 10 | 4041 | ||
BC000109.1 | - | 5 | 736 | |||
BC011722.2 | - | 16 | 2081 | 4300207 | ||
BC011761.2 | - | 16 | 2007 | 3622453 | ||
U61263.1 | - | 16 | 1989 |