Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTYMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165031.1 | NP_001158503.1 | 2 | 370 | ||
NM_001320902.1 | NP_001307831.1 | 2 | 370 | |||
NM_001320903.1 | NP_001307832.1 | 2 | 370 | |||
NM_001320904.1 | NP_001307833.1 | 2 | 370 | |||
NM_001320905.1 | NP_001307834.1 | 2 | 370 | |||
NM_012145.3 | NP_036277.2 | 2 | 370 | |||
NR_033255.1 | 2 | 370 | ||||
NR_135492.1 | 2 | 370 | ||||
GenBank mRNA | AA427388.1 | - | 2 | 111 | ||
AK223434.1 | - | 2 | 217 | |||
BC001827.2 | - | 2 | 210 | 3533327 | ||
BC147006.1 | - | 2 | 309 | 9056934 | ||
BC147010.1 | - | 2 | 309 | 9056938 | ||
BC171894.1 | - | 2 | 305 | |||
BC171902.1 | - | 2 | 309 | 9054556 | ||
BC171903.1 | - | 2 | 309 | |||
BG765702.1 | - | 2 | 116 | |||
BM552707.1 | - | 2 | 356 | |||
CN356277.1 | - | 2 | 206 | |||
CR541986.1 | - | 2 | 185 | |||
CR542015.1 | - | 2 | 185 | |||
CX166352.1 | - | 2 | 370 | |||
L16991.1 | - | 2 | 227 | |||
X54729.1 | - | 2 | 209 |