Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012467.3 | NP_036599.3 | 6 | 1048 | ||
XM_011522446.2 | XP_011520748.1 | 6 | 1787 | |||
XM_011522447.2 | XP_011520749.1 | 6 | 2661 | |||
GenBank mRNA | BE552396.1 | - | 1 | 310 |