Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127240.2 | NP_001120712.1 | 4 | 1317 | ||
NM_001127241.2 | NP_001120713.1 | 3 | 1028 | |||
NM_001127242.2 | NP_001120714.1 | 2 | 837 | |||
NM_014417.4 | NP_055232.1 | 4 | 1331 | |||
XM_006723141.3 | XP_006723204.1 | 4 | 1282 | |||
XM_011526722.2 | XP_011525024.1 | 4 | 1223 | |||
GenBank mRNA | AF332558.1 | - | 4 | 1332 | ||
AF354654.1 | - | 4 | 1319 | |||
AF354655.1 | - | 3 | 1030 | |||
AF354656.1 | - | 2 | 839 | |||
AK124984.1 | - | 1 | 1241 | |||
BG258126.1 | - | 1 | 101 | |||
U82987.1 | - | 4 | 1098 |