Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014788.3 | NP_055603.2 | 1, 2 | 380 | ||
NM_033219.2 | NP_150088.1 | 1, 2 | 380 | |||
XM_005252320.3 | XP_005252377.1 | 1, 2 | 445 | |||
XM_017015352.1 | XP_016870841.1 | 1, 2 | 445 | |||
XM_017015353.1 | XP_016870842.1 | 1, 2 | 445 | |||
XM_017015354.1 | XP_016870843.1 | 1, 2 | 436 | |||
GenBank mRNA | AF220130.1 | - | 1, 2 | 225 | ||
AF220131.1 | - | 1, 2 | 225 | |||
AK055833.1 | - | 2 | 253 | |||
AK292825.1 | - | 1, 2 | 247 | |||
BC006333.2 | - | 1, 2 | 233 | 4299815 | ||
BF797334.1 | - | 1, 2 | 338 | |||
D50919.1 | - | 1, 2 | 225 | |||
DC405577.1 | - | 1, 2 | 380 |