Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015956.2 | NP_057040.2 | 4 | 607 | ||
NM_146387.1 | NP_666499.1 | 5 | 471 | |||
NM_146388.1 | NP_666500.1 | 4 | 607 | |||
XM_011528045.2 | XP_011526347.1 | 4 | 602 | |||
GenBank mRNA | AB049635.1 | - | 4 | 322 | ||
AF132962.1 | - | 4 | 568 | |||
AY007152.1 | - | 3 | 260 | |||
BC000756.2 | - | 4 | 561 | 2819730 | ||
BC009858.2 | - | 4 | 348 | 3928448 | ||
BG758801.1 | - | 5 | 534 | |||
BM550378.1 | - | 5 | 469 |