Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016459.3 | NP_057543.2 | 4 | 522 | ||
GenBank mRNA | AF151024.1 | - | 4 | 601 | ||
AF338109.1 | - | 4 | 703 | |||
AK057549.1 | - | 3 | 1583 | |||
AK292706.1 | - | 4 | 497 | |||
BC009931.2 | - | 3 | 376 | 2989349 | ||
BC021275.2 | - | 4 | 503 | 4853250 | ||
BU601458.1 | - | 4 | 524 | |||
GQ477350.1 | - | 4 | 462 |