Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020186.2 | NP_064571.1 | 1 | 1238 | ||
GenBank mRNA | AF201933.1 | - | 1 | 127 | ||
BC020621.1 | - | 1 | 120 | 4766494 | ||
BC022858.1 | - | 1 | 128 | 4132451 | ||
BC028409.1 | - | 1 | 1246 |